Nalgene™ Safety Waste Funnel
Actual product may vary
Nalgene™ Safety Waste Funnel
Nalgene™ Safety Waste Funnel
Nalgene™ Safety Waste Funnel
Nalgene™ Safety Waste Funnel
Thermo Scientific
Nalgene™ Safety Waste Funnel
Contain waste temporarily on the benchtop or under the lab hood with these safety waste funnel systems. Funnels feature the easy snap safety latch to minimize spillage if accidentally tipped over and a larger diameter funnel for easy pouring.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure SizeDiameter (Metric) Opening
FIS10-437-3483B25.4 cm
FIS10-437-3338-43014 cm
Catalog number FIS10-437-34
Price (SGD)
1,222.00
Case
Closure Size:
83B
Diameter (Metric) Opening:
25.4 cm
Price (SGD)
1,222.00
Case
Temporarily contain waste on the benchtop or under the lab hood using Thermo Scientific™ Nalgene™ Safety Waste Funnel Systems. Funnels feature the Easy Snap Safety Latch to minimize spillage if accidentally tipped over and a larger diameter funnel for easy pouring. The vent system helps reduce fumes and splashing while the removable neck screen traps large particles, like stir bars. Designed for solvent, chemical and biological waste, it is recommended for use with Nalgene containers.
Features:
  • Polypropylene closure and funnel adaptor, polytetrafluoroethylene vent membrane
  • Available in 10 in. diameter funnel with 83B closure and 5.5 in. funnel with 38-430 closure
Includes:
  • HDPE funnel with attached lid
  • Polypropylene closure/funnel adapter
  • Screen insert
  • Polypropylene/PTFE 0.45μm vent plug
  • Filter
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
Closure Size83B
Closure TypeHDPE funnel with attached lid
DescriptionFunnel Safety Waste 10 in. HDPE/PP 83B Closure
Diameter (English) Outer10 in.
Length (Metric)5.6 cm
Quantity2/Cs.
Diameter (Metric) Opening25.4 cm
IncludesHDPE funnel with attached lid, polypropylene closure/funnel adapter, screen insert, polypropylene/PTFE 0.45µm vent plug, and filter.
Liner MaterialPTFE
For Use With (Application)For Safety Waste Systems
Length (English)2.2 in.
MaterialPolypropylene
Product TypeSafety Waste Funnel System
Unit SizeCase