PYREX™ Squibb Separatory Funnel with PTFE Standard Stopcock and Glass Standard Taper Stopper
Actual product may vary
PYREX™ Squibb Separatory Funnel with PTFE Standard Stopcock and Glass Standard Taper Stopper
Pear-shaped glass funnel with 65 mm stem length.
Have Questions?
Catalog NumberStopcockStopper NumberQuantity
FIS10-437-2F6; PTFE381/Cs.
Catalog number FIS10-437-2F
Price (SGD)
1,155.00
Each
Stopcock:
6; PTFE
Stopper Number:
38
Quantity:
1/Cs.
Price (SGD)
1,155.00
Each
The PTFE stopcock fits into the micro-finish barrel. The PTFE stopcock also reduces freezing problems and eliminates grease contamination.
For stopper only, see Cat. No. 7650.
Specifications
Diameter (Metric)Top16 cm
Capacity (Metric)2 L
MaterialBorosilicate Glass
For Use With (Application)For replacement glass standard taper stoppers
Length (Metric) Stem65 mm
Quantity1/Cs.
Stopcock6; PTFE
Stopper MaterialGlass
Stopper Number38
Product TypeSquibb Separatory Funnel
Unit SizeEach