Squibb Funnels with standard taper Stopcocks and Stoppers
Squibb Funnels with standard taper Stopcocks and Stoppers
Actual product may vary
Squibb Funnels with standard taper Stopcocks and Stoppers
Range of separatory funnels manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Have Questions?
Catalog NumberCapacity (Metric)Stopper Number
FIS10-436C500 mL27
Catalog number FIS10-436C
Price (SGD)
-
Capacity (Metric):
500 mL
Stopper Number:
27
  • Range of capacities from 125mL to 1000mL
  • Made entirely of borosilicate glass
  • Stem length: 75mm
Specifications
Capacity (Metric)500 mL
MaterialBorosilicate Glass
Height (Metric)323 mm
Certifications/ComplianceASTM E1096, Type IV
For Use With (Application)Eliminates contamination
Length (Metric) Stem75mm
QuantityEach
Stopper MaterialGlass
Stopper Number27
Product TypeSquibb Separatory Funnel
Unit SizeEA