Check Order Status
Short Stem Funnel, External Ridges
Actual product may vary
Bel-Art
Short Stem Funnel, External Ridges
Sturdy, polypropylene funnels with ridges help prevent vapor lock.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Height (Metric)
FIS10-371F
3845 mL
34.29 cm
FIS10-371E
720 mL
18.73 cm
FIS10-371C
180 mL
10.48 cm
FIS10-371D
480 mL
17.78 cm
4 Options
Catalog number
FIS10-371F
Price (SGD)
-
Capacity (Metric):
3845 mL
Height (Metric):
34.29 cm
Highly recommended for continuous laboratory work
Durable
Versatile
Steam autoclavable at 121°C (250°F)
Specifications
Diameter (Metric)Top
25.6 cm
Capacity (Metric)
3845 mL
Diameter (Metric) Stem
3.49 cm
Material
Polypropylene
Height (Metric)
34.29 cm
Autoclavable
Autoclavable
Length (Metric) Stem
13.97 cm
Product Type
Filling Funnel