PYREX™ Buchner Funnels with Fine Porosity Fritted Disc
Actual product may vary
PYREX™ Buchner Funnels with Fine Porosity Fritted Disc
Useful where filter paper would be attacked by material being filtered
Have Questions?
Catalog NumberCapacity (Metric)PorosityPore Size
FIS10-358-2G3000 mLMedium10 to 15 μm
Catalog number FIS10-358-2G
Price (SGD)
2,230.00
Each
Capacity (Metric):
3000 mL
Porosity:
Medium
Pore Size:
10 to 15 μm
Price (SGD)
2,230.00
Each
Useful where filter paper would be attacked by material being filtered
These funnels are especially useful where filter paper would be attacked by the material being filtered. The disc should not be subjected to pressure differential in excess of 1 atmosphere. The top edge is beaded.
Specifications
Capacity (Metric)3000 mL
Diameter (Metric) Stem25 mm
MaterialBorosilicate Glass
Height (Metric)355 mm
Certifications/ComplianceASTM E128
For Use With (Application)Useful where filter paper would be attacked by material being filtered
Diameter (Metric) Disc152 mm
IncludesFritted discs
Length (Metric) Stem110 mm
PorosityMedium
Pore Size10 to 15 μm
Product TypeBuchner Funnel
Unit SizeEach