Check Order Status
Large LDPE Funnels
Actual product may vary
Large LDPE Funnels
Large plastic funnels for transferring liquids at temperatures up to 80°C (180°F).
Have Questions?
Catalog Number
Diameter (Metric)Top
Height (Metric)
FIS10-349B
31.1cm
292.1 mm
Catalog number
FIS10-349B
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric)Top:
31.1cm
Height (Metric):
292.1 mm
Made of low-density polyethylene
External ribbing for air release
Internal ribbing in larger funnel
Withstand temperatures up to 80°C (180°F)
Stem length: 89mm
Specifications
Diameter (Metric)Top
31.1cm
Diameter (Metric) Stem
1.6cm
Material
Polyethylene
Height (Metric)
292.1 mm
For Use With (Application)
For transferring liquids
Length (Metric) Stem
8.9cm