Search Fisher Scientific
| Catalog Number | Diameter (Metric)Top | Diameter (Metric) Stem | Length (Metric) Stem | Height (Metric) |
|---|---|---|---|---|
| FIS10-346D | 12.5 cm | 2.5 cm | 2.5 cm | 13.2 cm |
| FIS10-346B | 7.5 cm | 1.7 cm | 1.7 cm | 9.4 cm |
| FIS10-346C | 10 cm | 2 cm | 2 cm | 12 cm |
These funnels have a short stem. For use in transferring powders or filling bottles.