PYREX™ 60° Angle Fluted Funnel, Short Stem
PYREX™ 60° Angle Fluted Funnel, Short Stem
Actual product may vary
PYREX™ 60° Angle Fluted Funnel, Short Stem
PYREX™ 60° Angle Fluted Funnel, Short Stem
PYREX™ 60° Angle Fluted Funnel, Short Stem
Useful in clinical and hospital labs; can be used in flasks or tubes without racks
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)TopDiameter (Metric) StemLength (Metric) StemHeight (Metric)
FIS10-329C7.5 cm0.8 cm7.5 cm13.9 cm
FIS10-329A5 cm0.8 cm6.5 cm10.8 cm
FIS10-329B6.5 cm0.8 cm6.5 cm12.1 cm
Catalog number FIS10-329C
Price (SGD)
671.00
PK
Add to cart
Diameter (Metric)Top:
7.5 cm
Diameter (Metric) Stem:
0.8 cm
Length (Metric) Stem:
7.5 cm
Height (Metric):
13.9 cm
Price (SGD)
671.00
PK
Add to cart
Supplied with short stems. Depressed inside flutings further decreases filtering time.
Specifications
Diameter (Metric)Top7.5 cm
Diameter (Metric) Stem0.8 cm
MaterialBorosilicate Glass
Height (Metric)13.9 cm
Angle60°
For Use With (Application)Useful in clinical and hospital labs, can be used in flasks or tubes without racks
Length (Metric) Stem7.5 cm
Product TypeFluted Filling Funnel
Unit SizePK