Kimble™ KIMAX™ Short Stem 58° Angle Addition Funnels
Kimble™ KIMAX™ Short Stem 58° Angle Addition Funnels
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Short Stem 58° Angle Addition Funnels
Bowls pressed to exact angle for fast filtering
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)TopDiameter (Metric) StemLength (Metric) StemCapacity (Metric)
FIS10-322F9cm0.9cm9.7cm150 mL
FIS10-322B4.5cm0.65cm5cm19 mL
FIS10-322D6.5cm0.8cm6.3cm60 mL
Catalog number FIS10-322F
Price (SGD)
310.00
PK
Add to cart
Diameter (Metric)Top:
9cm
Diameter (Metric) Stem:
0.9cm
Length (Metric) Stem:
9.7cm
Capacity (Metric):
150 mL
Price (SGD)
310.00
PK
Add to cart
Bowls pressed to exact angle for fast filtering
  • Heavy uniform walls
  • Beaded reinforced rims
  • Fire-polished ends
Specifications
Diameter (Metric)Top9cm
Capacity (Metric)150 mL
Diameter (Metric) Stem0.9cm
MaterialBorosilicate Glass
Certifications/ComplianceASTM E1095, Type I, Class A
Angle58°
Diameter (Metric) Filter15cm
For Use With (Application)Bowls pressed to exact angle for fast filtering
Length (Metric) Stem9.7cm
Unit SizePK