General-Purpose Extra-Long Forceps
Actual product may vary
General-Purpose Extra-Long Forceps
Fisherbrand General-Purpose Extra-Long Forceps are primarily used to remove specimens from deep bottles or jars.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
FIS10-316C30.5 cm.
FIS10-316A20.3 cm.
FIS10-316B25.4 cm.
Catalog number FIS10-316C
Price (SGD)
19.00
Each
Length (Metric):
30.5 cm.
Price (SGD)
19.00
Each
  • Fisherbrand™ General-Purpose Extra-Long Forceps are primarily used to remove specimens from deep bottles or jars
  • Our forceps are designed to exceed perfromance expectations and manufactured to exact standards -This is the Fisherbrand Advantage.
  • Grips specimens firmly
  • Helpful for hard-to-reach places
  • Useful for cleaning fish tanks
  • Useful in educational, research or medical setting
  • Perfect for everyday use
  • Built to last long
  • Guide pins allow round tips to meet precisely
  • Grooved handles provides user comfort and stability
  • Constructed of chrome
  • Hand polished to mirror finish
  • Electronically passivated for corrosion resistance
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Autoclavable up to 135°C
  • Latex-free
Specifications
Tip StyleRound, Serrated Jaws
DesignExtra-long tweezers
Length (English)12 in.
MaterialChrome
Length (Metric)30.5 cm.
Product TypeGeneral Purpose Forceps
Quantity (No. per Case)144
FinishHigh polish - mirror look
For Use With (Application)Multipurpose; Primarily used for to handle specimens in hard-to-reach areas
IncludesGuide pins
Unit SizeEach