Kimble™ Low Form Nessler Color Comparison Tubes
Actual product may vary
DWK Life Sciences
Kimble™ Low Form Nessler Color Comparison Tubes
Fill Kimble™ Low Form Nessler Color Comparison Tubes and let their shadowless bottom transmit undistorted light. No dark spots form when the tubes are filled with liquid and viewed from the top.
Have Questions?
Catalog NumberGraduations
FIS10-310-188100 mL
Catalog number FIS10-310-188
Price (SGD)
569.00
Case
Graduations:
100 mL
Price (SGD)
569.00
Case
Fill Kimble™ Low Form Nessler Color Comparison Tubes and let their shadowless bottom transmit undistorted light. No dark spots form when the tubes are filled with liquid and viewed from the top.
  • Durable white ceramic enamel rings and legend
  • Graduation Line
  • Clear
Specifications
SterilityNon-sterile
MaterialBorosilicate Glass
Volume (Metric)100 mL
ColorClear
AutoclavableAutoclavable
Length (Metric)200 mm
Diameter (Metric) Outer32 mm
Certifications/ComplianceUSP, ASTM E 438, APHA
Graduations100 mL
Scale TypeGraduated Ring
Length (English) Scale5.71 to 6.22 in.
QuantityUnmatched set of 6
Unit SizeCase