General-Purpose Student Forceps
General-Purpose Student Forceps
Actual product may vary
General-Purpose Student Forceps
Handle phosphorus, potassium and other materials with Fisherbrand™ General-Purpose Student Forceps. Perfectly designed for students in the laboratory setting.
Have Questions?
Catalog NumberLength (Metric)
FIS10-28012.7 cm
Catalog number FIS10-280
Price (SGD)
87.00
PK
Add to cart
Length (Metric):
12.7 cm
Price (SGD)
87.00
PK
Add to cart
Our instruments are designed to exceed performance expectations -This is the Fisherbrand Advantage.
  • Handles general lab materials
  • Perfect for students and researchers
  • Smooth handle grips
  • Durable and resists rusting
  • Made from corrosion-resistant ASTM™ standard stainless steel
  • Electronically passivated for corrosion resistance
  • Rockwell hardness (HRC): 40--45
  • Latex-free
  • Can be sterilized by Eto/Gamma radiation; Fully Autoclavable
  • Quality control per U.S. Government specs
  • Produced under ISO 9002, GMP, cGMP guidelines
Specifications
Tip StyleStraight
Length (English)5 in.
MaterialStainless Steel
Length (Metric)12.7 cm
Product TypeGeneral Purpose Forceps
Quantity (No. per Case)10 Pack
Unit SizePK