Kimble™ KIMAX™ Class A Cassia Flask
Kimble™ KIMAX™ Class A Cassia Flask
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Class A Cassia Flask
Calibrated To Contain
Have Questions?
Catalog number FIS10-245
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Calibrated To Contain
  • Official U.S. Pharmacopeia method to assay cinnamaldehyde and to analyze cassia, cinnamon, and clove extracts
  • Determine amount of unsaturated constituents in straw oil or similar wash oils, by sulfonation method
  • Tolerance: ±0.08mL
  • Capacity: 100 to 110mL in 0.1mL divisions
  • With standard taper 13 stopper
Specifications
CalibrationCalibrated To Contain
Capacity (Metric)110 mL
MaterialBorosilicate Glass
Height (English)10.23 in.
Height (Metric)26 cm
Tolerance±0.08 mL
GraduatedYes
Closure Size13
Unit SizePK