PYREX™ Low Actinic Class A Volumetric Flask with Glass Standard Taper Stopper
PYREX™ Low Actinic Class A Volumetric Flask with Glass Standard Taper Stopper
Actual product may vary
PYREX™ Low Actinic Class A Volumetric Flask with Glass Standard Taper Stopper
Calibrated To Contain
Have Questions?
Catalog NumberCapacity (Metric)Closure SizeClosure TypeTolerance
FIS10-227D250 mL16Hollow±0.12 mL
Catalog number FIS10-227D
Price (SGD)
1,100.00
EA
Add to cart
Capacity (Metric):
250 mL
Closure Size:
16
Closure Type:
Hollow
Tolerance:
±0.12 mL
Price (SGD)
1,100.00
EA
Add to cart
Low actinic stained glass provides protection for materials sensitive to light. The protective color is an integral part of the flask, which retains the mechanical strength, chemical stability, and thermal resistance of PYREX brand labware.
Specifications
Closure MaterialGlass
Diameter (English) Outer3 in.
Capacity (Metric)250 mL
MaterialGlass, Borosilicate Glass
Height (English)9.92 in.
Height (Metric)25.2 cm
Neck StyleLong
No. per Case12/Cs.
No. per Pack6/Pk.
Tolerance±0.12 mL
GraduatedYes
ClassA
Bottom ShapeFlat
Closure TypeHollow
Diameter (Metric) Outer78 mm
Closure Size16
Unit SizeEA