Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) |
---|---|
FIS10-211H | 2000 mL |
FIS10-211C | 100 mL |
FIS10-211B | 50 mL |
FIS10-211F | 500 mL |
FIS10-211G | 1 L |
Calibrated to Class A tolerances in accordance with ASTM E-542 and E-288. Each flask is individually serialized and supplied with a Certificate of Identification and Capacity, traceable to NIST standards. The graduation line is sharp and permanent; large white block letters make the inscription easy to read. Supplied with a standard taper stopper.
For stopper only, see Cat. No. 7650.
Tested and calibrated in an ISO/IEC 17025 certified laboratory.