Rtx™-5 Capillary Columns, 0.53 mm I.D.
Rtx™-5 Capillary Columns, 0.53 mm I.D.
Actual product may vary
DWK Life Sciences
Rtx™-5 Capillary Columns, 0.53 mm I.D.
General-Purpose Columns for Drugs, Solvent Impurities, Pesticides, Hydrocarbons, PCB Congeners, Essential Oils, Semivolatiles
Have Questions?
Catalog NumberLength (Metric)Film Thickness
FIS10-21030 m0.1 μm
Catalog number FIS10-210
Price (SGD)
-
Length (Metric):
30 m
Film Thickness:
0.1 μm
  • Removes all residual catalysts and low molecular weight fragments from the Rtx™-5 polymer
  • Provides a tight, unimodal distribution
  • Extremely low bleed
  • For Drugs, Solvent Impurities, Pesticides, Hydrocarbons, PCB Congeners (e.g., Aroclor Mixes), Essential Oils, Semivolatiles
Specifications
Diameter (Metric) Inner0.53 mm
Stationary PhaseRtx™-5
USP TypeG27, G36
Max. Temperature340°C
For Use With (Application)Drugs, Solvent Impurities, Pesticides, Hydrocarbons, PCB Congeners (e.g., Aroclor Mixes), Essential Oils, Semivolatiles
Temperature Range-60° to +320/340°C
Film Thickness0.1 μm
Length (Metric)30 m
PolarityLow
CoatingFused Silica
Quantity1/Pk.
Unit SizeCS