Reusable Glass Spinner Flasks
Reusable Glass Spinner Flasks
Actual product may vary
Reusable Glass Spinner Flasks
High-performance
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) OuterDiameter (Metric) Center Neck
FIS10-203B250 mL80 mm7 cm
FIS10-203A125 mL70 mm7 cm
Catalog number FIS10-203B
Price (SGD)
2,483.00
CS
Add to cart
Capacity (Metric):
250 mL
Diameter (Metric) Outer:
80 mm
Diameter (Metric) Center Neck:
7 cm
Price (SGD)
2,483.00
CS
Add to cart
  • Baffles enhance aeration and agitation of contents of the flask.
  • Unique impeller design ensures optimal stirring.
  • Sidearm designs permit easy access of 25 mL and 50 mL pipets.
Specifications
Closure ColorOrange
Closure MaterialPolypropylene
Diameter (English) Outer3.1 in.
Diameter (Metric) Center Cap7 cm
Diameter (Metric) Center Neck7 cm
Diameter (Metric) Inner Side Arm2.4 cm
Diameter (Metric) Side Arm3.2 cm
Diameter (Metric) Side Arm Cap3.2 cm
Capacity (Metric)250 mL
MaterialBorosilicate Glass
Height (English)7.2 in.
Height (Metric)18.5 cm
IncludesFlat center cap and 2 angled side arms
Neck StyleAngled
No. per Case1/Cs.
No. per Pack1/Pk.
Side Arm Angle45°
Side Arm Openings2
Thread SizeGL 32
SterilityNon-sterile
Closure TypeScrew Cap
Diameter (Metric) Outer80 mm
Unit SizeCS