Precision Clean™ II Ultra Clear Cleanroom Bags
Actual product may vary
Precision Clean™ II Ultra Clear Cleanroom Bags
Excellent clarity, available printed or unprinted
Have Questions?
Catalog NumberLength (English)Width (English)Thickness (English)
FIS10-200A36 in.24 in.6 mil
Catalog number FIS10-200A
Price (SGD)
56.00
Each
Length (English):
36 in.
Width (English):
24 in.
Thickness (English):
6 mil
Price (SGD)
56.00
Each
  • Ideal for packaging semiconductor wafer boxes, medical devices, cleanroom gowns, gloves, wipes or other products requiring a similar level of surface cleanliness
  • Can be sterilized by gamma radiation
  • Clear bags made from ultrapure resin
  • Manufactured and packaged in an ISO Class 4 cleanroom
  • Double-bagged with Certificate of Conformance label
Specifications
MaterialUltrapure Silicon-free Resin
Thickness (English)6 mil
Dimensions (L x W)36 x 24 in. (91.4 x 61cm)
Length (English)36 in.
Width (English)24 in.
Length (Metric)91.4 cm
Width (Metric)61 cm
Unit SizeEach