Cylindrical Dewar Flasks with Plastic Mesh Casing
Actual product may vary
Pope Scientific
Cylindrical Dewar Flasks with Plastic Mesh Casing
Pope Scientific Cylindrical Dewar Flasks with Plastic Mesh Casing, Depth Inside: 11.88 in., 30.2cm, Diameter Outer: 3.38 in., 8.6cm, Diameter Inner: 2.75 in., 7cm, Height: 13.26 in., 33.7cm, Capacity: 1L, Cap Lid Type: Plastic Mesh Cover, For Use With: Liquid nitrogen service, Borosilicate Glass
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDepth (English) InsideDiameter (English) OuterDiameter (English) InnerHeight (English)
FIS10-197D12 in.7 in.5.62 in.14.37 in.
FIS10-197B11.88 in.3.38 in.2.75 in.13.26 in.
Catalog number FIS10-197D
Price (SGD)
-
Depth (English) Inside:
12 in.
Diameter (English) Outer:
7 in.
Diameter (English) Inner:
5.62 in.
Height (English):
14.37 in.
Pope Scientific Cylindrical Dewar Flasks with Plastic Mesh Casing, Depth Inside: 11.88 in., 30.2cm, Diameter Outer: 3.38 in., 8.6cm, Diameter Inner: 2.75 in., 7cm, Height: 13.26 in., 33.7cm, Capacity: 1L, Cap Lid Type: Plastic Mesh Cover, For Use With: Liquid nitrogen service, Borosilicate Glass
Evacuated to 10-5 torr or better and covered by a protective mesh
  • Fully silvered double-walled borosilicate glass
  • Metal casing surrounds only bottom portion of flask
  • Protective plastic mesh covers remaining outer surface of glass
  • Special vented polyethylene stopper minimizes evaporation
  • Weighted bases for stability
  • No pads between walls on cylindrical Dewars
Specifications
Cap Lid TypePlastic Mesh Cover
Depth (English) Inside12 in.
Depth (Metric) Inside30.5cm
Diameter (English) Outer7 in.
Diameter (Metric) Outer17.8cm
For Use With (Application)Liquid nitrogen service
Height (English)14.37 in.
Height (Metric)36.5cm
ModelLab Grade
Capacity (Metric)4300 mL
Depth (Metric) Interior30.5 cm
Diameter (English) Inner5.62 in.
Diameter (Metric) Inner14.3 cm
MaterialBorosilicate Glass
StyleCylindrical, Wide Mouth
Unit SizeEach