▸
Cold Storage Products
▸
Insulated Containers
▸
Metal-Cased Cylindrical Dewar Flasks
Actual product may vary
Metal-Cased Cylindrical Dewar Flasks
Borosilicate glass with silvered double walls.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Includes
FIS10-195B
1 L
No handle
FIS10-195C
1900 mL
No handle
FIS10-196-11
1 L
Metal handle
3 Options
Catalog number
FIS10-195B
Price (SGD)
-
Capacity (Metric):
1 L
Includes:
No handle
Borosilicate glass with silvered double walls
For use with liquefied gases
Space between walls evacuated and sealed
Metal casing surrounds glass to protect user from accidental breakage
Special vented polyethylene stopper minimizes evaporation
Specifications
Cap Lid Type
Polyethylene Stopper
Diameter (English) Outer
3.38 in.
Diameter (Metric) Outer
8.6cm
For Use With (Application)
Liquefied gases
Height (English)
13.26 in.
Height (Metric)
33.7cm
Includes
No handle
Model
Lab Grade
Base Material
Aluminum
Capacity (Metric)
1 L
Diameter (English) Inner
2.76 in.
Diameter (Metric) Inner
7 cm
Material
Borosilicate Glass
Style
Cylindrical, Wide Mouth