Kimble™ KIMAX™ Filtering Flasks with Side Arm
Kimble™ KIMAX™ Filtering Flasks with Side Arm
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Filtering Flasks with Side Arm
Heavy walled with graduated scale, side tubulation, tooled neck
Have Questions?
Catalog NumberCapacity (Metric)
FIS10-181F1000 mL
Catalog number FIS10-181F
Price (SGD)
1,245.00
PK
Add to cart
Capacity (Metric):
1000 mL
Price (SGD)
1,245.00
PK
Add to cart
Heavy walled with graduated scale, side tubulation, tooled neck
  • Sizes 25, 50, and 125mL suitable for microanalysis
  • Sizes 25 and 500mL have side hose connections for 0.3125 in. I.D. (8mm) flexible tubing
  • Sizes 1000mL and larger accept 0.375 in. I.D. (9.5mm) tubing
Specifications
Certifications/ComplianceASTM E1406, Type II, Class I
Diameter (English) Outer5.3 in.
Capacity (Metric)1000 mL
MaterialBorosilicate Glass
Graduation Range300 to 1000 mL
Graduations50 mL
Height (English)9.37 in.
Height (Metric)23.8 cm
GraduatedYes
ClassClass I
Bottom ShapeFlat
Diameter (Metric) Outer13.5 cm
Closure Size8
Unit SizePK