PYREX™ Filtering Flasks with Sidearm Tubulation
PYREX™ Filtering Flasks with Sidearm Tubulation
Actual product may vary
PYREX™ Filtering Flasks with Sidearm Tubulation
Tubulations for easy connections to filter pumps
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Closure Size
FIS10-180F1 L23.8 cm8
FIS10-180G2000 mL29.2 cm9
FIS10-180H4 L35.6 cm12
FIS10-180C125 mL11 cm5
FIS10-180D250 mL14.6 cm6
Catalog number FIS10-180F
Price (SGD)
1,171.00
Pack
Add to cart
Capacity (Metric):
1 L
Height (Metric):
23.8 cm
Closure Size:
8
Price (SGD)
1,171.00
Pack
Add to cart
Tubulations for easy connections to filter pumps
These flasks, with tubulation, are blown in special molds, in shapes designed to give maximum mechanical strength. For convenience, the flasks are graduated to show approximate capacity. The neck finish affords an excellent fit for rubber stoppers. Tubulation O.D. on sizes up to 1000 mL is approximately 10 mm. Tubulation O.D. for 2000 mL, and 4000 mL is approximately 12.5 mm.
Specifications
DescriptionStyle: Filtering; Top style: Heavy duty rim
Diameter (English) Outer5 in.
Diameter (Metric) Outer Tubing1 cm
Capacity (Metric)1 L
MaterialBorosilicate Glass
Graduation Range300 to 1000 mL
Graduations50 mL
Height (English)9.37 in.
Height (Metric)23.8 cm
Neck StyleNarrow Mouth
No. per Case12/Cs.
No. per Pack6/Pk.
DisposableReusable
Mouth StyleNarrow
SterilityNon-sterile
Diameter (Metric) Outer127 mm
Closure Size8
Unit SizePack