BioCoat™ 96-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate
BioCoat™ 96-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate
Actual product may vary
BioCoat™ 96-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: With Lid
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackaging
FIS10-129120/Pk., 80/Cs.
FIS08-774-1275/Cs
Catalog number FIS10-1291
Price (SGD)
3,959.00
CS
Add to cart
Packaging:
20/Pk., 80/Cs.
Price (SGD)
3,959.00
CS
Add to cart
Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.
Applications include:
  • Attachment and spreading of a variety of cell types
  • Cell differentiation and neurite outgrowth
  • Attachment of fastidious transfected cell lines
  • Support survival of primary neurons in culture
  • Serum-free or reduced-serum culture
Source
  • PDL, synthetic (MW 75-150 kD)
  • PLL, synthetic (MW 30-70 kD)
Quality Control
  • Tested for ability to promote firm attachment of RCG cells
  • Tested and found negative for bacteria and fungi
Storage
  • 4°C to 30°C under dry conditions.
  • Specifications
    Certifications/ComplianceANSI, SBS
    FormatStandard
    LidWith Lid
    Array8 x 12
    Packaging20/Pk., 80/Cs.
    ColorClear
    Product TypeStandard
    MaterialPolystyrene
    No. of Wells96
    SterilitySterile
    Surface TreatmentPoly-D-Lysine
    Well ShapeRound, Flat Bottom (F-well)
    For Use With (Application)Attachment and spreading of a variety of cell types, cell differentiation and neurite outgrowth, attachment of fastidious transfected cell lines, support survival of primary neurons in culture, serum-free or reduced serum culture
    No. per Case80/Cs.
    No. per Pack20/Pk.
    Quantity80/Cs.
    Shelf Life6 Months
    SourcePoly-D-Lysine
    Well ColorClear
    Unit SizeCS
    Contents & Storage
    4°C to 30°C