Two- and Three-Litre Polycarbonate Erlenmeyer Flasks
Actual product may vary
Two- and Three-Litre Polycarbonate Erlenmeyer Flasks
Manufactured from optically clear polycarbonate for optimum visibility and durability. Corning™ Two-and Three-Liter Disposable Polycarbonate Erlenmeyer Flasks are ideal for shaker cultures applications.
Have Questions?
Catalog NumberCapacity (Metric)Bottom Shape
FIS10-126-432000 mLBaffled
Catalog number FIS10-126-43
Price (SGD)
900.00
Case
Capacity (Metric):
2000 mL
Bottom Shape:
Baffled
Price (SGD)
900.00
Case
  • Fernbach design increases surface area for gas exchange
  • Made from optically-clear polycarbonate
  • Ideal for shaker culture applications
  • Baffled bottoms improve mixing and increase gas exchange
  • Vent caps supplied in every case of product for applications requiring sterile gas exchange
  • Sterilized by gamma radiation and nonpyrogenic
  • RNase- and DNase-free
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifications
Capacity (English)67.6 oz.
Closure ColorOrange
Closure MaterialPolypropylene
DescriptionTop style: Threaded
Capacity (Metric)2000 mL
MaterialPolycarbonate
For Use With (Application)For shaker culture applications
Graduations100 mL
Neck StyleWide Mouth
No. per Case6/Cs.
No. per Pack1/Pk.
DisposableSingle-use
Mouth StyleWide
SterilitySterile
Bottom ShapeBaffled
Closure TypeVented
Unit SizeCase