PYREX™ Long Neck Kjeldahl Flasks
PYREX™ Long Neck Kjeldahl Flasks
Actual product may vary
PYREX™ Long Neck Kjeldahl Flasks
Long necks, round bottoms, substantial walls
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS10-110E500 mL
FIS10-110D300 mL
Catalog number FIS10-110E
Price (SGD)
-
Capacity (Metric):
500 mL
Of all the common laboratory test methods, the Kjeldahl method for determining nitrogen subjects the boiling flask to the most severe thermal and chemical abuse. These Kjeldahl flasks, specially styled, have tooled finished necks for extra strength and uniform stopper fit. Wall thickness is controlled to offer the optimum balance between thermal shock and chemical resistance. Can be used in ASTM E-258 procedure. Care must be taken to ensure the bottoms of the flasks do not come in contact with the heating elements. Most Kjeldahl equipment manufacturers have ring supports for this purpose.
Reference: ASTM E-1377.
Specifications
DescriptionTop style: Tooled
Diameter (English) Outer3.97 in.
Capacity (Metric)500 mL
MaterialBorosilicate Glass
Height (English)12.7 in.
Height (Metric)32.4 cm
Neck StyleLong
No. per Case24/Cs.
No. per Pack6/Pk.
DisposableReusable
Bottom ShapeRound
Diameter (Metric) Outer101 mm
Closure Size6
Unit SizePK