Kimble™ KIMAX™ Flasks with Pennyhead standard taper Stopper
Kimble™ KIMAX™ Flasks with Pennyhead standard taper Stopper
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Flasks with Pennyhead standard taper Stopper
For volatile or hygroscopic liquids
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS10-100F500 mL
FIS10-100C50 mL
Catalog number FIS10-100F
Price (SGD)
315.00
PK
Add to cart
Capacity (Metric):
500 mL
Price (SGD)
315.00
PK
Add to cart
  • Narrow mouths
  • Graduated scales
Specifications
Closure MaterialGlass
DescriptionCapacity scale
Diameter (English) Outer4.01 in.
Capacity (Metric)500 mL
MaterialBorosilicate Glass
Graduation Range100 to 500 mL
Graduations50 mL
Height (English)6.25 in.
Height (Metric)15.9 cm
Mouth StyleNarrow
Closure TypePenny Stopper
Diameter (Metric) Outer102 mm
Closure Size32
Unit SizePK