PYREX™ Narrow Mouth Erlenmeyer Flask with Standard Taper Stopper
PYREX™ Narrow Mouth Erlenmeyer Flask with Standard Taper Stopper
Actual product may vary
PYREX™ Narrow Mouth Erlenmeyer Flask with Standard Taper Stopper
Narrow mouths
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Material
FIS10-098G500 mLBorosilicate Glass
FIS10-098D125 mLBorosilicate Glass
Catalog number FIS10-098G
Price (SGD)
164.00
EA
Add to cart
Capacity (Metric):
500 mL
Material:
Borosilicate Glass
Price (SGD)
164.00
EA
Add to cart
For convenience, these flasks are graduated to show approximate capacity. The 25 mL, 50 mL, and 125 mL flasks are satisfactory for use as a weighing bottle.
For stopper only, see Cat. No. 7650.
Specifications
Diameter (English) Outer3.97 in.
Capacity (Metric)500 mL
MaterialBorosilicate Glass
Graduation Range100 to 500 mL
Graduations50 mL
Height (English)8.46 in.
Height (Metric)21.5 cm
Neck StyleNarrow Mouth
No. per Case8/Cs.
No. per Pack1/Pk.
Writing SurfaceYes
DisposableReusable
Mouth StyleNarrow
SterilityNon-sterile
Closure TypeBarrel Head
Diameter (Metric) Outer101 mm
Closure Size32
Unit SizeEA