Kimble™ KIMAX™ Iodine Determination Flasks with PTFE Stopper
Kimble™ KIMAX™ Iodine Determination Flasks with PTFE Stopper
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Iodine Determination Flasks with PTFE Stopper
For analysis of dry shellac, varnishes, oils, fatty acids
Have Questions?
Catalog NumberCapacity (Metric)
FIS10-096A125 mL
Catalog number FIS10-096A
Price (SGD)
393.00
PK
Add to cart
Capacity (Metric):
125 mL
Price (SGD)
393.00
PK
Add to cart
For analysis of dry shellac, varnishes, oils, fatty acids
250mL size
  • Analyze dry shellac and shellac varnishes according to ASTM™ D 29
  • Determine total iodine value of oils, fatty acids, and bodied oils according to ASTM D 1541
500mL size
  • Test drying oils for use in paints and varnishes according to ASTM D 555
Specifications
Closure MaterialPTFE
Diameter (English) Outer2.59 in.
Capacity (Metric)125 mL
MaterialBorosilicate Glass
Height (English)4.9 in.
Height (Metric)12.4 cm
Writing SurfaceMatte Finish Ceramic Enamel Marking
Diameter (Metric) Outer66 mm
Closure Size22
Unit SizePK