PYREX™ Wide Mouth Heavy-Duty Erlenmeyer Flasks
PYREX™ Wide Mouth Heavy-Duty Erlenmeyer Flasks
Actual product may vary
PYREX™ Wide Mouth Heavy-Duty Erlenmeyer Flasks
Use these flasks for a variety of applications. PYREX™ Wide Neck Heavy-Duty Erlenmeyer Flasks are designed for longer life and feature a high resistance to chemical attack, rugged tooled top finish and wide mouth.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Diameter (Metric) OuterGraduation Range
FIS10-090E2000 mL37.2 cm198 mm400 to 1800 mL
FIS10-090B250 mL18.4 cm106 mm50 to 200 mL
FIS10-090C500 mL23.8 cm137 mm100 to 500 mL
FIS10-090A125 mL14.6 cm84 mm50 to 125 mL
FKS122014081 L29.2 cm168 mm250 to 1000 mL
FKS106420912000 mL37.2 cm198 mm400 to 1800 mL
Catalog number FIS10-090E
Price (SGD)
115.00
EA
Add to cart
Capacity (Metric):
2000 mL
Height (Metric):
37.2 cm
Diameter (Metric) Outer:
198 mm
Graduation Range:
400 to 1800 mL
Price (SGD)
115.00
EA
Add to cart
Use these flasks for a variety of applications. PYREX™ Wide Neck Heavy-Duty Erlenmeyer Flasks are designed for longer life and feature a high resistance to chemical attack, rugged tooled top finish and wide mouth.
A wide mouth flask especially recommended for use as a titration flask. For convenience, these flasks are graduated to show the approximate capacity and have an extra large marking spot.
Specifications
DescriptionWhite Ceramic Enamel scale graduations; Top style: Heavy duty rim
Diameter (English) Outer7.79 in.
Capacity (Metric)2000 mL
MaterialBorosilicate Glass, Borosilicate Glass
Graduation Range400 to 1800 mL
Graduations100 mL
Height (English)14.6 in.
Height (Metric)37.2 cm
Neck StyleWide Mouth
No. per Case8/Cs.
No. per Pack1/Pk.
ShapeConical
Writing SurfaceExtra Large White Square Marking
DisposableReusable
Mouth StyleWide
SterilityNon-sterile
Diameter (Metric) Outer198 mm
Closure Size13
Unit SizeEA