PYREX™ Narrow Mouth Erlenmeyer Flask, Standard Taper Joint
PYREX™ Narrow Mouth Erlenmeyer Flask, Standard Taper Joint
Actual product may vary
PYREX™ Narrow Mouth Erlenmeyer Flask, Standard Taper Joint
Narrow Mouth
Have Questions?
Catalog NumberCapacity (Metric)Standard Taper Joint
FIS10-047E500 mL24/40
Catalog number FIS10-047E
Price (SGD)
645.00
PK
Add to cart
Capacity (Metric):
500 mL
Standard Taper Joint:
24/40
Price (SGD)
645.00
PK
Add to cart
A range of Erlenmeyer flasks with joints. For convenience, these flasks are graduated to show approximate capacity.
Cat. No. 5000-500: Reference ASTM E-123.
Specifications
DescriptionTop style: Standard taper joint
Diameter (English) Outer3.97 in.
Capacity (Metric)500 mL
MaterialBorosilicate Glass, Borosilicate Glass
Graduation Range100 to 500 mL
Graduations50 mL
Height (English)9.17 in.
Height (Metric)23.3 cm
Neck StyleNarrow Mouth
No. per Case12/Cs.
No. per Pack6/Pk.
Standard Taper Joint24/40
Writing SurfaceExtra Large Marking
DisposableReusable
Mouth StyleNarrow
SterilityNon-sterile
Diameter (Metric) Outer101 mm
Unit SizePK