PYREX™ Narrow Mouth Erlenmeyer Culture Flask with Baffles
Actual product may vary
PYREX™ Narrow Mouth Erlenmeyer Culture Flask with Baffles
Usable with shakers—three baffles provide optimal and consistent agitation
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS10-041-5A125 mL
FIS10-041-5B250 mL
Catalog number FIS10-041-5A
Price (SGD)
849.00
Case
Capacity (Metric):
125 mL
Price (SGD)
849.00
Case
Heavy tooled rim for maximum durability. The same baffle design as found on our Delong flasks promotes optimal and consistent solution agitation when used with flask shakers.
Specifications
DescriptionTop style: Tooled
Diameter (English) Outer2.7 in.
Capacity (Metric)125 mL
MaterialBorosilicate Glass
For Use With (Application)Ideal for shaker cultures
Height (English)6.1 in.
Height (Metric)15.5 cm
Neck StyleNarrow Mouth
No. per Case6/Cs.
No. per Pack6/Pk.
Mouth StyleNarrow
SterilityNon-sterile
Bottom ShapeBaffled
Diameter (Metric) Outer70 mm
Closure Size5
Unit SizeCase