Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Includes |
---|---|---|
FIS10-040Q | 6 L | No Stopper |
FIS10-040C | 50 mL | No Stopper |
FIS10-040D | 125 mL | No Stopper |
FIS10-040F | 250 mL | No Stopper |
FIS10-040G | 300 mL | No Stopper |
FIS10-040P | 4 L | No Stopper |