PYREX™ Long Neck Flat Bottom Flasks, Tooled Mouth
PYREX™ Long Neck Flat Bottom Flasks, Tooled Mouth
Actual product may vary
PYREX™ Long Neck Flat Bottom Flasks, Tooled Mouth
Usual globe-shaped Florence boiling flasks
Have Questions?
Catalog NumberCapacity (Metric)
FIS10-035J6 L
Catalog number FIS10-035J
Price (SGD)
538.00
CS
Add to cart
Capacity (Metric):
6 L
Price (SGD)
538.00
CS
Add to cart
The 12,000 mL size has a short neck.
Specifications
DescriptionTop style: Tooled
Diameter (English) Outer9.3 in.
Capacity (Metric)6 L
MaterialBorosilicate Glass
Height (English)14.4 in.
Height (Metric)36.7 cm
Neck StyleLong
No. per Case1/Cs.
No. per Pack1/Pk.
DisposableReusable
Bottom ShapeFlat
Diameter (Metric) Outer237 mm
Closure Size11
Unit SizeCS