Whatman™ Reeve Angel 802 Prepleated Qualitative Circles
Whatman™ Reeve Angel 802 Prepleated Qualitative Circles
Actual product may vary
Fisherbrand
Whatman™ Reeve Angel 802 Prepleated Qualitative Circles
Coarse retention
Have Questions?
Catalog NumberDiameter (Metric)
FIS09-901B15 cm
Catalog number FIS09-901B
Price (SGD)
153.00
PK
Add to cart
Diameter (Metric):
15 cm
Price (SGD)
153.00
PK
Add to cart
  • Folded for fast filtration of coarse particles and gelatinous precipitates
  • Qualitative Grade; nominal retention 30-35μm; 0.26mm thick Pleating and creped surface increase effective area, improving both filtering speed and extending filter life
  • Wet strengthened†
Specifications
Description802 15 cm 100/pk
FormatDisc
Particle Retention15 μm
Pore Size30 to 35 μm
For Use With (Application)A prepleated filter for use with a conical filter funnel, offers fast filtration and high loading capacity for analysis involving coarse particles or gelatinous precipitates.
Grade802
PorosityCoarse
Quantity100/Pk.
Thickness (Metric)0.26 mm
Dimensions (L x W)15.00 cm
Diameter (Metric)15 cm
Unit SizePK