▸
Dishes
▸
Weighing Dishes and Papers
▸
Low-Nitrogen Weighing Paper
Actual product may vary
Fisherbrand
Low-Nitrogen Weighing Paper
For analytical balances
Have Questions?
Change view
Catalog Number
Dimensions (L x W)
FIS09-898-12B
4 x 4 in. (100 x 100mm)
FIS09-898-12A
3 x 3 in. (76 x 76mm)
FIS09-898-12C
6 x 6 in. (152 x 152mm)
3 Options
Catalog number
FIS09-898-12B
Price (SGD)
21.00
Special offer
Ends:
31-Mar-2026
33.00
Save 12.00 (36%)
Pack
Add to cart
Dimensions (L x W):
4 x 4 in. (100 x 100mm)
Price (SGD)
21.00
Special offer
Ends:
31-Mar-2026
33.00
Save 12.00 (36%)
Pack
Add to cart
For analytical balances
Sufficient body to support sample
Smooth textured to avoid trapping material
Does not absorb moisture
Specifications
Dimensions (L x W)
4 x 4 in. (100 x 100mm)
For Use With (Application)
For analytical balances
Length (English)
4 in.
Length (Metric)
100 mm
Width (English)
4 in.
Width (Metric)
100 mm
Product Type
Low Nitrogen Paper
Shape
Square