Qualitative Grade Plain Filter Paper Circles - P5 Grade
Qualitative Grade Plain Filter Paper Circles - P5 Grade
Actual product may vary
Fisherbrand
Qualitative Grade Plain Filter Paper Circles - P5 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FIS09-801J4.25 cm
FIS09-801B9 cm
FIS09-801A7 cm
FIS09-801E15 cm
FIS09-801D12.5 cm
Catalog number FIS09-801J
Price (SGD)
11.00
PK
Add to cart
Diameter (Metric):
4.25 cm
Price (SGD)
11.00
PK
Add to cart
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
  • Cellulose fiber papers
  • Good wet strength
  • Suitable for gravity, vacuum filtration systems and low suction filtration
  • Particle Retention: 5–10μm
  • Medium porosity
  • Filtration Speed: Herzberg, 230 seconds
  • Flowrate: Slow—60mL/min.
  • Surface: Smooth
Not suitable for nitrogen analysis.
Applications:
General lab uses: clinical and spot testing, clarifying pharmaceuticals.
Specifications
FormatDisc
MaterialCellulose
Particle Retention5 to 10 μm
Filtration Speed230 sec.
Flow RateSlow: 60mL/min.
For Use With (Application)General lab uses: clinical and spot testing, clarifying pharmaceuticals
GradeP5
PorosityMedium
Quantity100/Pk.
Diameter (Metric)4.25 cm
Unit SizePK