Qualitative Grade Plain Filter Paper Circles - P8 Grade
Qualitative Grade Plain Filter Paper Circles - P8 Grade
Actual product may vary
Fisherbrand
Qualitative Grade Plain Filter Paper Circles - P8 Grade
Ideal for clarifying and removing particles of 20μm and larger, and in applications that do not require a low-ash or ashless paper.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FIS09-795K33 cm
FIS09-795C9 cm
FIS09-795E12.5 cm
FIS09-795A5.5 cm
FIS09-795F15 cm
FIS09-795H20.5 cm
Catalog number FIS09-795K
Price (SGD)
70.00
PK
Add to cart
Diameter (Metric):
33 cm
Price (SGD)
70.00
PK
Add to cart
  • Cellulose fiber papers
  • Good wet strength
  • Suitable for gravity, vacuum filtration systems and low suction filtration
  • Particle Retention: 20–25μm
  • Coarse porosity
  • Filtration Speed: Herzberg, 55 seconds
  • Flowrate: Fast—160mL/min.
  • Surface: Smooth, open texture
Not suitable for nitrogen analysis.
Applications:
Suitable for coarse and gelatinous precipitates. Retains Fe(OH)3, Al(OH)3, metallic sulfides, iron ores. For clarifying solutions, oils. For zinc analysis (Fe ppt removal). Size 19cm (09-804) widely used for sugar analysis.
Specifications
FormatDisc
MaterialCellulose
Particle Retention20 to 25 μm
Filtration Speed55 sec.
Flow RateFast: 160mL/min.
For Use With (Application)Suitable for coarse and gelatinous precipitates. Retains Fe(OH)3, Al(OH)3, metallic sulfides, iron ores. For clarifying solutions, oils. For zinc analysis (Fe ppt removal). Size 19cm (09-804) widely used for sugar analysis.
GradeP8
PorosityCoarse
Quantity100/Pk.
Diameter (Metric)33 cm
Unit SizePK