Search Thermo Fisher Scientific
| Catalog Number | Diameter (Metric) |
|---|---|
| FIS09-795H | 20.5 cm |
| FIS09-795C | 9 cm |
| FIS09-795E | 12.5 cm |
| FIS09-795A | 5.5 cm |
| FIS09-795K | 33 cm |
| FIS09-795F | 15 cm |
Not suitable for nitrogen analysis.
Applications:
Suitable for coarse and gelatinous precipitates. Retains Fe(OH)3, Al(OH)3, metallic sulfides, iron ores. For clarifying solutions, oils. For zinc analysis (Fe ppt removal). Size 19cm (09-804) widely used for sugar analysis.