Use as replacement receivers for Corning Disposable Sterile Filter Systems or as storage bottles. Corning™ Disposable Sterile Bottles are easy to grip and ideal for storage of media, buffers and other aqueous solutions.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FIS09-761-4
250 mL
FIS09-761-140
150 mL
CRG430281
250 mL
FIS09-761-10
500 mL
FKS10278951
500 mL
FIS09-761-11
1 L
FKS10124291
1 L
7 Options
Catalog number FIS09-761-4
Price (SGD)
379.00
Case
Capacity (Metric):
250 mL
Price (SGD)
379.00
Case
Disposable polystyrene bottles for storage of media, buffers, and other aqueous solutions
Corning low profile style has easy grip sides that facilitate handling
Plug seal caps (45 mm) provide an airtight seal and help minimize the risk of contamination.
Bottles can be used with Corning vacuum filtration systems with 45 mm neck sizes (Cat. Nos. 430626, 430627, 431161, 430049, 430513, 430514,431118, 430015, and 431174).
Sterile
Nonpyrogenic
Specifications
Capacity (Metric)250 mL
For Use With (Application)Applications: Storage of media, buffers and other aqueous solutions