Check Order Status
Glass Microanalysis Vacuum Filter Holders
Actual product may vary
Glass Microanalysis Vacuum Filter Holders
Designed for use with Fisherbrand vacuum filters.
Have Questions?
Catalog Number
Material
FIS09-753E
Silicone, Fritted Glass
Catalog number
FIS09-753E
Price (SGD)
461.00
EA
Add to cart
Material:
Silicone, Fritted Glass
Price (SGD)
461.00
EA
Add to cart
Designed for use with Fisherbrand vacuum filters.
Fritted glass support
Graduated top funnel
Specifications
For Use With (Equipment)
Fisherbrand Vacuum Filter
Material
Silicone, Fritted Glass
Filtration Area
2.1 cm
2
Type
Vacuum Filter Holder
Volume (Metric)
25 mL
Diameter (English) Inside
0.98 in.
Diameter (Metric) Inside
25 mm