Glass Microanalysis Vacuum Filter Holders
Glass Microanalysis Vacuum Filter Holders
Actual product may vary
Glass Microanalysis Vacuum Filter Holders
Glass Microanalysis Vacuum Filter Holders
Glass Microanalysis Vacuum Filter Holders
Glass Microanalysis Vacuum Filter Holders
Designed for use with Fisherbrand vacuum filters.
Have Questions?
Catalog NumberType
FIS09-753C13 mL/100 mL Funnel
Catalog number FIS09-753C
Price (SGD)
-
Type:
13 mL/100 mL Funnel
  • Fritted glass support
  • Graduated top funnel
Specifications
MaterialSilicone, Fritted Glass, Anodized Aluminum
Filtration Area1.1 cm2
Type13 mL/100 mL Funnel
Volume (Metric)100 mL
Unit SizeEA