Nalgene™ Reusable Filter Funnel with Clamp
Nalgene™ Reusable Filter Funnel with Clamp
Actual product may vary
Nalgene™ Reusable Filter Funnel with Clamp
Nalgene™ Reusable Filter Funnel with Clamp
Nalgene™ Reusable Filter Funnel with Clamp
Thermo Scientific
Nalgene™ Reusable Filter Funnel with Clamp
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
Have Questions?
Catalog NumberCapacity (Metric)
FIS09-747250 mL
Catalog number FIS09-747
Price (SGD)
841.00
CS
Add to cart
Capacity (Metric):
250 mL
Price (SGD)
841.00
CS
Add to cart
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Specifications
Diameter (Metric)Top47 mm
Capacity (Metric)250 mL
MaterialPSU
AutoclavableAutoclavable
For Use With (Application)High-throughput particle and microbiological QC testing
IncludesRugged Polysulfone Funnel, Polypropylene Copolymer Cover with 3 ports, Friction-fit TPE Caps (3 caps), Support Plate for 47 mm membrane, Silicone Gasket, No. 8 Stopper with 0.5625 in. hole, Aluminum Clamp
Quantity1 Ea.
Product TypeFiltering Funnel
Unit SizeCS