PYREX™ Extractor System with Soxhlet Extractor and Allihn Condenser
PYREX™ Extractor System with Soxhlet Extractor and Allihn Condenser
Actual product may vary
PYREX™ Extractor System with Soxhlet Extractor and Allihn Condenser
Complete unit
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)Height (English)Standard Taper Joint BottomStandard Taper Joint Top
FIS09-556D33.81 oz.34.64 in.29/4271/60
FIS09-556C16.9 oz.31.61 in.24/4055/50
FIS09-556A4.22 oz.25.78 in.24/4034/45
FIS09-556B8.45 oz.27.99 in.24/4045/50
Catalog number FIS09-556D
Price (SGD)
-
Capacity (English):
33.81 oz.
Height (English):
34.64 in.
Standard Taper Joint Bottom:
29/42
Standard Taper Joint Top:
71/60
This apparatus consists of a Soxhlet extractor, an Allihn condenser, and a flask, all which are equipped with standard taper joints. The tubulation O.D. on the condenser is 10 mm.
For extractor only, see Cat. No. 3740.
For flask only on sizes 125 mL through 500 mL, see Cat. No. 4100.
For other replacement parts: see flask, Cat. No. 3840-1 LFO and condensers, Cat. Nos. 3840-SCO, 3840-MCO, 3840-LCO, 3840-XLCO.
Specifications
Product TypeSoxhlet Extraction Apparatus
Capacity (Metric)1 L
Capacity (English)33.81 oz.
IncludesAllihn condenser
Standard Taper Joint Bottom29/42
Standard Taper Joint Top71/60
Diameter (English) Outer Tube0.39 in.
Diameter (Metric) Outer Tube1 cm
Height (English)34.64 in.
Height (Metric)88 cm
Length (English) Tube12.55 in.
Length (Metric) Tube31.9 cm
Unit SizeEA