Kimble™ KIMAX™ Short Neck Flask with standard taper Joints
Kimble™ KIMAX™ Short Neck Flask with standard taper Joints
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Short Neck Flask with standard taper Joints
Flat Bottom Boiling
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS09-552C500 mL
FIS09-552A125 mL
Catalog number FIS09-552C
Price (SGD)
132.00
EA
Add to cart
Capacity (Metric):
500 mL
Price (SGD)
132.00
EA
Add to cart
  • Freestanding flasks with standard taper 24/40 joints
  • Designed from ASTM™ E 1403, Type I, Class 4 requirements
Specifications
Diameter (English) Outer4 in.
Capacity (Metric)500 mL
MaterialBorosilicate Glass
Height (English)6.1 in.
Height (Metric)15.5 cm
Neck StyleShort
Standard Taper Joint24/40
Diameter (Metric) Outer102 mm
Unit SizeEA