Kimble™ KIMAX™ Soxhlet Apparatus with Allihn Condenser, Complete unit
Kimble™ KIMAX™ Soxhlet Apparatus with Allihn Condenser, Complete unit
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Soxhlet Apparatus with Allihn Condenser, Complete unit
Designed from Fed. Spec. NNN-F-240
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)Height (English)Length (English) TubeStandard Taper Joint Top
FIS09-551C16.9 oz.28.34 in.1.96 in.55/50
FIS09-551B8.45 oz.25.98 in.1.57 in.45/50
Catalog number FIS09-551C
Price (SGD)
2,185.00
EA
Add to cart
Capacity (English):
16.9 oz.
Height (English):
28.34 in.
Length (English) Tube:
1.96 in.
Standard Taper Joint Top:
55/50
Price (SGD)
2,185.00
EA
Add to cart
  • Complete assembly includes 09-551-10 Series Allihn condenser, 09-553 Series Soxhlet extraction tube with standard taper 24/40 joint at bottom, 09-552 Series flask
  • All connected by interchangeable ground joints
  • Hose connections on condenser accept 0.31 in. I.D. (8mm) flexible tubing
Specifications
Product TypeExtraction Apparatus Assembly
For Use With (Equipment)Paper Thimble (43 dia. x 123 mm H)
Capacity (Metric)500 mL
Capacity (English)16.9 oz.
IncludesAllihn condenser, soxhlet extraction tube
Standard Taper Joint Top55/50
Height (English)28.34 in.
Height (Metric)72 cm
Length (English) Tube1.96 in.
Length (Metric) Tube5 cm
Unit SizeEA