Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems
Actual product may vary
Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems
Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems
Thermo Scientific
Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems
Achieve superior flexibility in a small footprint with our B2 mini gel electrophoresis systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
FIS09-528-110BEasyCast
B2EasyCast
MICB-2EasyCast
FIS09-528-118EasyCast, Buffer Exchange Ports
Catalog number FIS09-528-110B
Price (SGD)
-
Includes:
EasyCast
Achieve superior flexibility in a small footprint with Thermo Scientific™ Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems. The simple-to-use casting method, as well as multiple comb and tray options, make this product Owl's most popular system.
Features:
  • Two comb slots on the U.V. Transmissible (UVT) gel tray double sample capacity by doubling the number of sample wells
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands
Ordering Information:
Available with optional buffer exchange ports (B2-BP) for use with an external recirculation pump
Includes:
  • Buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • EasyCast gasketed U.V. Transmissible (UVT) gel tray
  • Two combs: 12 & 20 well, double-sided, 1.0/1.5 mm thick
Specifications
Certifications/ComplianceCE marked
Gel SizeMini
Product LineOwl
TypeMini Gel Electrophoresis System
IncludesEasyCast
Sample Size8 to 48
Volume (Metric) Buffer800 mL
Dimensions (L x W x H)9.63 x 7.06 x 3.75 in. (24.5 x 18 x 9.5 cm)
Length (English)9.63 in.
Length (Metric)24.5 cm
Length (Metric) Gel14 cm
Width (English)7.06 in.
Width (Metric)18 cm
Width (Metric) Gel12 cm
Height (English)3.75 in.
Height (Metric)9.5 cm
Max. Voltage150 V
Unit SizeEach