▸
Buffers and Standards
▸
Electrochemistry Standards
▸
Conductivity and TDS Standards
▸
Conductivity Calibration Standards, YSI™
Element Search
Structure Search
YSI
Conductivity Calibration Standards, YSI™
NIST-traceable conductivity solutions to calibrate both field instruments and lab instruments and cells together for maximum accuracy
Have Questions?
Catalog Number
Quantity
Packaging
Conductivity
FIS09-390-21
8 x 1 pt.
Plastic Bottle
1,000μS/cm (Fresh water)
Catalog number
FIS09-390-21
Price (SGD)
564.00
Case
Add to cart
Quantity:
8 x 1 pt.
Packaging:
Plastic Bottle
Conductivity:
1,000μS/cm (Fresh water)
Request bulk or custom format
Price (SGD)
564.00
Case
Add to cart
Specifications
Packaging
Plastic Bottle
Traceability to NIST
Yes
Analyte or Component Names
Fresh Water Conductivity Calibrator
Density
1g/mL
Conductivity
1,000μS/cm (Fresh water)
View more
Conductivity solutions are inspected with reference to primary standard solutions according to OIML recommendation 56
Bottles include a table of corrections at temperatures between 20 and 30°C
Available as: 1 Quart or in a box of 8 individual pints