▸
Buffers and Standards
▸
Electrochemistry Standards
▸
Conductivity and TDS Standards
▸
Conductivity Calibration Standards, YSI™
Element Search
Structure Search
YSI
Conductivity Calibration Standards, YSI™
NIST-traceable conductivity solutions to calibrate both field instruments and lab instruments and cells together for maximum accuracy
Have Questions?
Change view
Quantity:
1 qt.
8 x 1 pt.
Packaging:
View all
Glass Bottle
Conductivity:
View all
100,000μS/cm
Catalog number
FIS09-390-20
Price (SGD)
324.00
Each
Add to cart
Quantity:
1 qt.
Packaging:
Glass Bottle
Conductivity:
100,000μS/cm
Request bulk or custom format
Price (SGD)
324.00
Each
Add to cart
Specifications
Packaging
Glass Bottle
Conductivity
100,000μS/cm
Type
Conductivity Calibration Standard
Quantity
1 qt.
Unit Size
Each
Conductivity solutions are inspected with reference to primary standard solutions according to OIML recommendation 56
Bottles include a table of corrections at temperatures between 20 and 30°C
Available as: 1 Quart or in a box of 8 individual pints