Drierite™ Gas Purifier
Drierite™ Gas Purifier
Actual product may vary
Drierite™ Gas Purifier
Drierite™ Gas Purifier
Drierite™ Gas Purifier
Drierite™ Gas Purifier
Designed for gas chromatography and other applications requiring dry, pure gas. W.A. Hammond Drierite™ Gas Purifier dries, purifies, and filters gases used for chromatography and spectrometry.
Have Questions?
Catalog number FIS09-206
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Designed for gas chromatography and other applications requiring dry, pure gas. W.A. Hammond Drierite™ Gas Purifier dries, purifies, and filters gases used for chromatography and spectrometry.
  • Specially designed for gas chromatography and other applications requiring dry, pure gas
  • Used in gas chromatography to prevent ghost peaks and to improve baselines 
  • Used for carrier gases and for flame ionization detector air and hydrogen gases
  • Used to remove moisture and impurities from gases when purging spectrophotometer compartments
  • dries gases to a dryness of 0.005mg/L
  • Color changes from blue to pink upon exhaustion
  • Removes impurities that have an effective molecular diameter of less than 5 angstroms(A)
Specifications
Diameter (Metric) Outer6.65 cm
Product TypeGas Purifier
MaterialPolycarbonate
For Use With (Application)For use in gas chromatography to prevent ghost peaks and to improve baselines, also used to remove moisture and impurities from gases when purging spectrophotometer compartments
Height (Metric)28.7 cm
Height (English)11.3 in.
Includes15 cu. in. each of indicating drierite absorbent and 5A molecular sieves
Diameter (English) Outer2.62 in.
Unit SizeEA