Search Thermo Fisher Scientific
Catalog Number | For Use With (Equipment) | Resin Type | Capacity | For Use With (Application) |
---|---|---|---|---|
FIS09-034-10 | Bantam™ Deionizer, B-Pure™ | Strong Cation Exchange | 3170 grains | Cation Removal |
D0760 | Bantam™ Deionizer, B-Pure™ | Strong Anion Exchange | 1680 grains | Anion Removal |
FIS09-034-92 | Bantam™ Deionizer, B-Pure™ | Strong Anion Exchange | 1680 grains | Anion Removal |
D0809 | Bantam™ Deionizer, B-Pure™ | Mixed Bed | 875 grains | |
FIS09-034-3 | Bantam™ Deionizer, B-Pure™ | Mixed Bed | 875 grains | |
D0811 | B-Pure™ | Porous Strong Ion Exchange | 30 g | Oxygen Removal |
D0813 | Bantam™ Deionizer, B-Pure™ | Activated Carbon | 5000 gal. | Organic Removal |
FIS09-034-5 | Bantam™ Deionizer, B-Pure™ | Activated Carbon | 5000 gal. | Organic Removal |
D0815 | Bantam™ Deionizer, B-Pure™ | Strong Cation Exchange | 3170 grains | Cation Removal |
D0832 | Bantam™ Deionizer, B-Pure™ | Mixed Bed, Activated Carbon | 785 grains/1000 gal. | Ultrapure and Organic Removal |
FIS09-028-33 | Bantam™ Deionizer, B-Pure™ | Mixed Bed, Activated Carbon | 785 grains/1000 gal. | Ultrapure and Organic Removal |
D0836 | B-Pure™ | Macroreticular, Activated Carbon | 2000 gal. | Pretreatment Macropure |